Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAB31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006868.3 | NP_006859.2 | 7 | 2963 | ||
XM_017025528.1 | XP_016881017.1 | 8 | 3345 | |||
XM_017025529.1 | XP_016881018.1 | 8 | 3333 | |||
GenBank mRNA | AF183421.1 | - | 7 | 2920 | ||
CN429637.1 | - | 1 | 70 |