Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BCL2L12 SIRNA Assays ›
Gene Symbol:
BCL2L12
BCL2L12P1
Gene Name:
BCL2 like 12
BCL2 like 12 pseudogene 1
Gene Aliases:
-
-
Chromosome Location:
Chr.19: 49665072 - 49673916 on Build GRCh38
Chromosome Location:
Chr.3: 131526468 - 131527141 on Build GRCh38
UniGene ID:
Hs.289052
Hs.640518
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
120719
Validated
Purification
In-Vivo Ready
Standard
HPLC
Size
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
1 umol
5 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
10 umol
Availability
Inventoried
Catalog #
AM51331
Price
6,301.00
Your Price
Online offer:
6301.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BCL2L12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040668.1
NP_001035758.1
4
1195
NM_001282516.1
NP_001269445.1
4
1198
NM_001282517.1
NP_001269446.1
3
1055
NM_001282519.1
NP_001269448.1
4
1198
NM_001282520.1
NP_001269449.1
4
1198
NM_138639.1
NP_619580.1
4
1198
NR_104200.1
3
1055
NR_104201.1
4
1198
NR_104202.1
3
1055
NR_104203.1
3
1055
NR_104204.1
3
1055
XM_017027345.1
XP_016882834.1
4
1265
XM_017027346.1
XP_016882835.1
3
1125
GenBank mRNA
BC007724.2
-
4
333
BC104004.1
-
4
771
40024338
BC104005.1
-
4
774
40024342
BC104006.1
-
4
774
40024345
BI159786.1
-
3
221
FJ868798.1
-
4
516
FJ868799.1
-
3
373
FJ868800.1
-
4
516
FJ868801.1
-
3
373
FJ868802.1
-
4
516
FJ868803.1
-
3
373
FJ911906.1
-
4
516
HM347053.1
-
3
373
KR820244.1
-
2
1352
KR820245.1
-
2
1352
KR820246.1
-
2
1352
KR820247.1
-
2
1352
KR820248.1
-
2
1352
KR820249.1
-
2
1401
KR820250.1
-
2
1401
KR820251.1
-
2
1401
KR820252.1
-
2
1401
KR820253.1
-
2
1401
KR820254.1
-
3
1133
KR820255.1
-
3
1133
KR820256.1
-
3
1133
KR820257.1
-
3
1133
KR820258.1
-
3
1182
KR820259.1
-
3
1182
KR820260.1
-
3
1182
KR820261.1
-
3
1182
KR820262.1
-
3
1182
KR820263.1
-
3
1130
KR820264.1
-
3
1130
KR820265.1
-
3
1130
KR820266.1
-
3
1179
KR820267.1
-
3
1179
KR820268.1
-
3
1179
KR820269.1
-
3
1179
KR820270.1
-
3
735
KR820271.1
-
3
735
KR820272.1
-
3
735
KR820273.1
-
3
735
KR820274.1
-
3
784
KR820275.1
-
3
784
KR820276.1
-
3
784
KR820277.1
-
3
784
KR820278.1
-
4
565
KR820279.1
-
4
565
KR820280.1
-
4
565
KR820281.1
-
4
565
KR820282.1
-
4
513
KR820283.1
-
4
513
KR820284.1
-
4
562
KR820285.1
-
4
562
KR820286.1
-
4
562
KR820287.1
-
4
562
KR820288.1
-
2
990
KR820289.1
-
2
990
KR820290.1
-
2
990
KR820291.1
-
2
990
KR820292.1
-
2
990
Gene Symbol: BCL2L12P1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
GQ202621.1
-
1
345
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00369327_m1
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
positive regulation of transcription from RNA polymerase II promoter
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of cellular senescence
p53 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0