Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIG1 SIRNA Assays ›
          Gene Symbol:
          LIG1
          Gene Name:
          DNA ligase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 48115445 - 48170603 on Build GRCh38
          UniGene ID:
          Hs.1770
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 121248

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LIG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000234.2 NP_000225.1 10, 11 1286
          NM_001289063.1 NP_001275992.1 9, 10 1193
          NM_001289064.1 NP_001275993.1 8, 9 1082
          NM_001320970.1 NP_001307899.1 10, 11 1283
          NM_001320971.1 NP_001307900.1 9, 10 1196
          NR_110296.1 10, 11 1283
          NR_135497.1 10, 11 1286
          NR_135499.1 9, 10 1196
          NR_135500.1 10, 11 1463
          NR_135501.1 9, 10 1193
          XM_017026809.1 XP_016882298.1 8, 9 815
          GenBank mRNA AB208791.1 - 10, 11 1006
          AK225863.1 - 11, 12 1022
          AK297296.1 - 8, 9 793
          AK300370.1 - 9, 10 934
          AK300607.1 - 5, 6 622
          AK303644.1 - 9, 10 909
          AK314210.1 - 10, 11 993
          BC108318.1 - 10, 11 1286 6141535
          BC110622.2 - 9, 10 935 40035687
          M36067.1 - 10, 11 986

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via recombination
          DNA metabolic process
          DNA repair
          transcription-coupled nucleotide-excision repair
          base-excision repair
          nucleotide-excision repair, DNA gap filling
          mismatch repair
          anatomical structure morphogenesis
          V(D)J recombination
          DNA ligation involved in DNA repair
          cell division
          DNA biosynthetic process
          Okazaki fragment processing involved in mitotic DNA replication
          DNA binding
          DNA ligase activity
          DNA ligase (ATP) activity
          ATP binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0