Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ANGPT1 SIRNA Assays ›
          Gene Symbol:
          ANGPT1
          Gene Name:
          angiopoietin 1
          Gene Aliases:
          AGP1, AGPT, ANG1
          Chromosome Location:
          Chr.8: 107249482 - 107498055 on Build GRCh38
          UniGene ID:
          Hs.369675
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 121279

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ANGPT1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146.4 NP_001137.2 1 607
          NM_001199859.2 NP_001186788.1 1 607
          GenBank mRNA AB084454.1 - 1 110
          AK304717.1 - 1 60
          AY121504.1 - 1 110
          AY124380.1 - 1 110
          BC152411.1 - 1 461 8862514
          BC152419.1 - 1 473 8862529
          BX648814.1 - 1 608
          D13628.2 - 1 461
          U83508.1 - 1 419

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_009640, NM_001286062, AK012027, AK088439, AK172868, and others.

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          in utero embryonic development
          negative regulation of protein phosphorylation
          regulation of endothelial cell proliferation
          sprouting angiogenesis
          positive regulation of receptor internalization
          negative regulation of cytokine secretion involved in immune response
          negative regulation of cell adhesion
          activation of transmembrane receptor protein tyrosine kinase activity
          positive regulation of endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          regulation of skeletal muscle satellite cell proliferation
          hemopoiesis
          cell differentiation
          heparin biosynthetic process
          positive regulation of protein ubiquitination
          cell-substrate adhesion
          regulation of tumor necrosis factor production
          positive regulation of peptidyl-serine phosphorylation
          protein localization to cell surface
          negative regulation of protein import into nucleus
          negative regulation of apoptotic process
          negative regulation of vascular permeability
          regulation of I-kappaB kinase/NF-kappaB signaling
          regulation of protein binding
          negative regulation of neuron apoptotic process
          positive regulation of blood vessel endothelial cell migration
          positive regulation of GTPase activity
          positive regulation of cell adhesion
          Tie signaling pathway
          positive regulation of peptidyl-tyrosine phosphorylation
          leukocyte migration
          positive chemotaxis
          positive regulation of protein kinase B signaling
          positive regulation of ERK1 and ERK2 cascade
          glomerulus vasculature development
          negative regulation of endothelial cell apoptotic process
          regulation of macrophage migration inhibitory factor signaling pathway
          Ras guanyl-nucleotide exchange factor activity
          receptor tyrosine kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-kbcv7:80/100.66.79.224:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline