Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PSMC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199163.1 | NP_001186092.1 | 8, 9 | 1094 | ||
NM_002805.5 | NP_002796.4 | 8, 9 | 959 | |||
GenBank mRNA | AF035309.1 | - | 1380 | |||
AK095836.1 | - | 7, 8 | 2193 | |||
AK098484.1 | - | 2744 | ||||
AK290758.1 | - | 8, 9 | 1057 | |||
AK291878.1 | - | 8, 9 | 931 | |||
BC001932.1 | - | 8, 9 | 910 | 2820259 | ||
BC002367.2 | - | 8, 9 | 889 | 2821416 | ||
D44467.1 | - | 8, 9 | 889 | |||
L38810.1 | - | 8, 9 | 896 |