Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNT10B SIRNA Assays ›
          Gene Symbol:
          WNT10B
          Gene Name:
          Wnt family member 10B
          Gene Aliases:
          SHFM6, WNT-12
          Chromosome Location:
          Chr.12: 48965340 - 48971858 on Build GRCh38
          UniGene ID:
          Hs.91985
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 121440

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNT10B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003394.3 NP_003385.2 4 1128
          XM_011538722.1 XP_011537024.1 3 526
          XM_011538724.1 XP_011537026.1 5 901
          XM_017019919.1 XP_016875408.1 2 1160
          GenBank mRNA AB070724.1 - 3 732
          AK312906.1 - 4 1132
          BC096353.3 - 4 768 40010101
          BC096354.1 - 4 768 40010102
          BC096355.3 - 5 626
          BC096356.1 - 4 768 40010104
          U81787.1 - 4 1047
          X97057.1 - 2 554

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          chondrocyte differentiation
          lipid metabolic process
          cell cycle arrest
          smoothened signaling pathway
          positive regulation of cell proliferation
          positive regulation of G2/M transition of mitotic cell cycle
          myoblast differentiation involved in skeletal muscle regeneration
          Wnt signaling pathway
          neuron differentiation
          positive regulation of bone mineralization
          positive regulation of epithelial cell differentiation
          regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of apoptotic process
          cell fate commitment
          negative regulation of fat cell differentiation
          positive regulation of osteoblast differentiation
          positive regulation of RNA polymerase II transcriptional preinitiation complex assembly
          regulation of skeletal muscle tissue development
          skeletal muscle fiber development
          negative regulation of epithelial cell proliferation
          protein stabilization
          sensory perception of taste
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of anagen
          canonical Wnt signaling pathway
          bone trabecula formation
          fungiform papilla development
          cellular response to retinoic acid
          cellular response to cAMP
          cellular response to parathyroid hormone stimulus
          hematopoietic stem cell proliferation
          cellular response to hydrostatic pressure
          positive regulation of canonical Wnt signaling pathway
          frizzled binding
          receptor agonist activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0