Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BCL10 SIRNA Assays ›
          Gene Symbol:
          BCL10
          Gene Name:
          B-cell CLL/lymphoma 10
          Gene Aliases:
          CARMEN, CIPER, CLAP, IMD37, c-E10, mE10
          Chromosome Location:
          Chr.1: 85265776 - 85276904 on Build GRCh38
          UniGene ID:
          Hs.193516
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 121470

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BCL10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001320715.1 NP_001307644.1 2 792
          NM_003921.4 NP_003912.1 2 792
          XM_011542397.2 XP_011540699.1 2 661
          XM_011542398.2 XP_011540700.1 2 666
          XM_011542399.2 XP_011540701.1 2 814
          GenBank mRNA AF057700.1 - 2 486
          AF082283.1 - 3 946
          AF100338.1 - 2 487
          AF105066.1 - 2 240
          AF127386.1 - 2 240
          AF134395.1 - 2 485
          AJ006288.1 - 3 930
          AK291346.1 - 2 450
          BC053617.1 - 2 362 6143423
          EF189176.1 - 2 345

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          B cell apoptotic process
          neural tube closure
          stimulatory C-type lectin receptor signaling pathway
          toll-like receptor signaling pathway
          response to molecule of bacterial origin
          adaptive immune response
          negative regulation of mature B cell apoptotic process
          cellular defense response
          I-kappaB kinase/NF-kappaB signaling
          cell death
          response to fungus
          immunoglobulin mediated immune response
          positive regulation of protein ubiquitination
          response to food
          positive regulation of mast cell cytokine production
          Fc-epsilon receptor signaling pathway
          lymphotoxin A biosynthetic process
          interleukin-6 biosynthetic process
          positive regulation of phosphorylation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          innate immune response
          positive regulation of interleukin-8 biosynthetic process
          positive regulation of transcription, DNA-templated
          T cell receptor signaling pathway
          regulation of T cell receptor signaling pathway
          positive regulation of T cell activation
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          protein homooligomerization
          protein heterooligomerization
          T cell apoptotic process
          cellular response to mechanical stimulus
          positive regulation of extrinsic apoptotic signaling pathway
          protease binding
          transcription coactivator activity
          protein binding
          protein C-terminus binding
          transcription factor binding
          kinase activator activity
          enzyme binding
          kinase binding
          protein kinase binding
          ubiquitin protein ligase binding
          identical protein binding
          ubiquitin binding
          protein kinase B binding
          protein self-association
          NF-kappaB binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qwrcb:80/100.66.75.127:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0