Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320715.1 | NP_001307644.1 | 2 | 802 | ||
NM_003921.4 | NP_003912.1 | 2 | 802 | |||
XM_011542397.2 | XP_011540699.1 | 2 | 671 | |||
XM_011542398.2 | XP_011540700.1 | 2 | 676 | |||
XM_011542399.2 | XP_011540701.1 | 2 | 824 | |||
GenBank mRNA | AF057700.1 | - | 2 | 496 | ||
AF082283.1 | - | 3 | 956 | |||
AF100338.1 | - | 2 | 497 | |||
AF105066.1 | - | 2 | 250 | |||
AF127386.1 | - | 2 | 250 | |||
AF134395.1 | - | 2 | 495 | |||
AJ006288.1 | - | 3 | 940 | |||
AK291346.1 | - | 2 | 460 | |||
BC053617.1 | - | 2 | 372 | 6143423 | ||
EF189176.1 | - | 2 | 355 |