Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NUPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042483.1 | NP_001035948.1 | 3 | 864 | ||
NM_012385.2 | NP_036517.1 | 3 | 810 | |||
GenBank mRNA | AF069073.1 | - | 3 | 645 | ||
AF135266.2 | - | 3 | 601 | |||
BC002434.2 | - | 3 | 590 | 3347264 |