Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PORCN SIRNA Assays ›
          Gene Symbol:
          PORCN
          Gene Name:
          porcupine homolog (Drosophila)
          Gene Aliases:
          DHOF, FODH, MG61, PORC, PPN
          Chromosome Location:
          Chr.X: 48508954 - 48520814 on Build GRCh38
          UniGene ID:
          Hs.386453
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 122143

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PORCN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282167.1 NP_001269096.1 8 1220
          NM_022825.3 NP_073736.2 8 1075
          NM_203473.2 NP_982299.1 9 1093
          NM_203474.1 NP_982300.1 8 945
          NM_203475.2 NP_982301.1 10 1108
          XM_005272635.2 XP_005272692.1 9 1448
          XM_005272636.2 XP_005272693.1 8 1430
          XM_005272637.1 XP_005272694.1 11 1390
          XM_006724545.3 XP_006724608.1 11 1173
          XM_006724546.3 XP_006724609.1 11 1245
          XM_006724547.1 XP_006724610.1 10 965
          XM_011543948.2 XP_011542250.1 8 1359
          XM_017029735.1 XP_016885224.1 10 1473
          XM_017029736.1 XP_016885225.1 8 932
          GenBank mRNA AF317058.1 - 9 929
          AF317059.1 - 8 914
          AF317060.1 - 8 911
          AF317061.1 - 7 896
          AK056340.1 - 8 1402
          AK057727.1 - 7 1322
          AK293347.1 - 8 1185
          AK314745.1 - 9 978
          AK316378.1 - 7 967
          BC019080.2 - 9 1040 4559533
          L08239.1 - 8 845

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001173355, XM_006256716, XM_006256717, BC158642.
          This siRNA also targets the following transcripts in Mouse: BU707440, BC032284, AK010405, AB036748, AB036749, and others.

          Panther Classification:

          Molecular Function -

          acetyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein lipidation
          glycoprotein metabolic process
          Wnt signaling pathway
          protein palmitoleylation
          canonical Wnt signaling pathway
          Wnt-protein binding
          palmitoleoyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0