Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NUPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042483.1 | NP_001035948.1 | 2 | 518 | ||
NM_012385.2 | NP_036517.1 | 2 | 464 | |||
GenBank mRNA | AF069073.1 | - | 2 | 299 | ||
AF135266.2 | - | 2 | 255 | |||
AK312135.1 | - | 2 | 247 | |||
BC002434.2 | - | 2 | 244 | 3347264 | ||
BF965595.1 | - | 2 | 300 | |||
BP214401.1 | - | 2 | 464 | |||
BT006896.1 | - | 2 | 197 | |||
CR542144.1 | - | 2 | 197 |