Search
Search

ANGPTL4| Gene Symbol: ANGPTL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139314.2 | NP_647475.1 | 4 | 799 | ||
| XM_005272484.3 | XP_005272541.1 | 4 | 797 | |||
| GenBank mRNA | AB056477.1 | - | 4 | 773 | ||
| AF153606.1 | - | 4 | 754 | |||
| AF169312.1 | - | 4 | 668 | |||
| AF202636.1 | - | 4 | 776 | |||
| AK222489.1 | - | 4 | 771 | |||
| AK222524.1 | - | 4 | 769 | |||
| AK303269.1 | - | 5 | 700 | |||
| AY358275.1 | - | 4 | 767 | |||
| BC023647.2 | - | 4 | 743 | 5088323 | ||