Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXN SIRNA Assays ›
Gene Symbol:
PXN
Gene Name:
paxillin
Gene Aliases:
-
Chromosome Location:
Chr.12: 120210439 - 120265771 on Build GRCh38
UniGene ID:
Hs.446336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
12253
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080855.2
NP_001074324.1
11
1548
NM_001243756.1
NP_001230685.1
10
1590
NM_002859.3
NP_002850.2
10
1446
NM_025157.4
NP_079433.3
10
1570
XM_005253917.3
XP_005253974.1
6
945
XM_006719532.1
XP_006719595.1
14
3131
XM_006719534.1
XP_006719597.1
14
3286
XM_006719535.3
XP_006719598.1
15
3214
XM_006719536.2
XP_006719599.1
14
3103
XM_011538622.1
XP_011536924.1
15
3213
XM_011538623.1
XP_011536925.1
15
3400
XM_017019726.1
XP_016875215.1
14
3699
XM_017019727.1
XP_016875216.1
14
3499
XM_017019728.1
XP_016875217.1
14
3490
XM_017019729.1
XP_016875218.1
14
3555
XM_017019730.1
XP_016875219.1
14
3699
XM_017019731.1
XP_016875220.1
15
3584
XM_017019733.1
XP_016875222.1
12
3114
XM_017019734.1
XP_016875223.1
14
3011
XM_017019735.1
XP_016875224.1
11
2868
XM_017019736.1
XP_016875225.1
12
2538
XM_017019737.1
XP_016875226.1
12
2394
XM_017019738.1
XP_016875227.1
11
2229
XM_017019739.1
XP_016875228.1
10
2127
XM_017019740.1
XP_016875229.1
11
2085
XM_017019741.1
XP_016875230.1
11
1876
XM_017019742.1
XP_016875231.1
10
1983
XM_017019743.1
XP_016875232.1
11
1684
GenBank mRNA
AB209034.1
-
13
3224
AK122964.1
-
11
1876
AK128712.1
-
8
1896
AK299486.1
-
8
877
AK314204.1
-
10
1405
BC003647.2
-
2
100
BC136787.1
-
10
1380
9020777
BC136794.1
-
10
1380
9020784
BC144410.1
-
11
1482
9052931
BX648777.1
-
10
1564
D86862.1
-
10
1406
D86863.1
-
9
1448
U14588.1
-
10
1378
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell adhesion
cell-matrix adhesion
signal transduction
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
cellular response to reactive oxygen species
vascular endothelial growth factor receptor signaling pathway
growth hormone receptor signaling pathway
integrin binding
protein binding
beta-catenin binding
zinc ion binding
vinculin binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qwrcb:80/100.66.75.127:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0