Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NOS1AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164757.1 | NP_001158229.1 | 6 | 840 | ||
NM_014697.2 | NP_055512.1 | 6 | 855 | |||
GenBank mRNA | AB007933.2 | - | 6 | 855 | ||
BC041382.2 | - | 6 | 857 | |||
BC112295.1 | - | 6 | 556 | 8327763 | ||
BC143771.1 | - | 6 | 541 | 9052285 | ||
BC143780.1 | - | 6 | 541 |