Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H11A SIRNA Assays ›
Gene Symbol:
ZC3H11A
LOC441155
Gene Name:
zinc finger CCCH-type containing 11A
zinc finger CCCH-type domain-containing-like
Gene Aliases:
ZC3HDC11A
-
Chromosome Location:
Chr.1: 203795537 - 203854128 on Build GRCh38
Chromosome Location:
Chr.6: 65301418 - 65306069 on Build GRCh38
UniGene ID:
Hs.532399
Hs.546686
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
122718
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H11A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319238.1
NP_001306167.1
9
1669
NM_001319239.1
NP_001306168.1
9
1564
NM_014827.5
NP_055642.3
9
1240
XM_005245643.2
XP_005245700.1
9
1259
XM_005245644.3
XP_005245701.1
8
943
XM_006711673.2
XP_006711736.1
9
1106
XM_011510199.1
XP_011508501.1
7
2311
XM_011510200.1
XP_011508502.1
8
1331
XM_011510201.1
XP_011508503.1
9
1517
XM_011510202.1
XP_011508504.1
7
1256
XM_017002956.1
XP_016858445.1
8
2565
XM_017002957.1
XP_016858446.1
8
2417
XM_017002958.1
XP_016858447.1
8
2404
XM_017002959.1
XP_016858448.1
9
1477
XM_017002960.1
XP_016858449.1
8
2565
XM_017002961.1
XP_016858450.1
7
2311
XM_017002962.1
XP_016858451.1
8
1331
XM_017002963.1
XP_016858452.1
9
1123
XM_017002964.1
XP_016858453.1
7
2311
XM_017002965.1
XP_016858454.1
6
1640
GenBank mRNA
AB014563.1
-
6
751
AK095858.1
-
9
1538
AK296983.1
-
5
598
BC014268.2
-
9
1078
3844154
BC038513.2
-
9
1395
BX648271.1
-
6
4053
BX649148.1
-
6
4855
CR627439.1
-
7
2261
CR627446.1
-
6
4364
Gene Symbol: LOC441155
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271675.1
NP_001258604.1
2
1272
XM_017010889.1
XP_016866378.1
2
788
GenBank mRNA
AK302514.1
-
2
1228
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA processing factor
Gene Ontology Categories:
Function(s)
Process(es)
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
poly(A)+ mRNA export from nucleus
mRNA 3'-end processing
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-bfkrn:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0