Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PPP4R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122964.2 | NP_001116436.2 | 17 | 3888 | ||
NM_001282850.1 | NP_001269779.1 | 16 | 3792 | |||
NM_020463.3 | NP_065196.1 | 15 | 3633 | |||
XM_005264442.2 | XP_005264499.1 | 17 | 3867 | |||
XM_005264444.2 | XP_005264501.1 | 16 | 3729 | |||
XM_005264445.2 | XP_005264502.1 | 16 | 3708 | |||
XM_005264446.2 | XP_005264503.1 | 15 | 3612 | |||
XM_005264447.2 | XP_005264504.1 | 16 | 3264 | |||
XM_017004531.1 | XP_016860020.1 | 16 | 3771 | |||
XM_017004532.1 | XP_016860021.1 | 16 | 3243 | |||
XM_017004533.1 | XP_016860022.1 | 15 | 3168 | |||
XM_017004534.1 | XP_016860023.1 | 15 | 3147 | |||
XM_017004535.1 | XP_016860024.1 | 14 | 3009 | |||
XM_017004536.1 | XP_016860025.1 | 14 | 2988 | |||
XM_017004537.1 | XP_016860026.1 | 18 | 4079 | |||
XM_017004538.1 | XP_016860027.1 | 18 | 3991 | |||
XM_017004539.1 | XP_016860028.1 | 17 | 3875 | |||
XM_017004540.1 | XP_016860029.1 | 17 | 3874 | |||
GenBank mRNA | AB037808.1 | - | 17 | 3751 | ||
AK023148.1 | - | 6 | 1781 | |||
AK026437.1 | - | 12 | 2890 | |||
AK098321.1 | - | 1 | 1707 | |||
AL122106.1 | - | 2 | 1086 | |||
AL136556.1 | - | 7 | 1881 | |||
BC060855.1 | - | 15 | 3522 | 5259789 |