Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130028.1 | NP_001123500.1 | 2 | 981 | ||
NM_003992.4 | NP_003983.2 | 2 | 131 | |||
XM_005254151.3 | XP_005254208.1 | 2 | 981 | |||
XM_011521205.2 | XP_011519507.1 | 2 | 981 | |||
XM_011521206.2 | XP_011519508.1 | 2 | 981 | |||
XM_011521209.2 | XP_011519511.2 | 2 | 1032 | |||
XM_017021905.1 | XP_016877394.1 | 2 | 1032 | |||
XM_017021906.1 | XP_016877395.1 | 2 | 1032 | |||
XM_017021907.1 | XP_016877396.1 | 2 | 1032 | |||
XM_017021908.1 | XP_016877397.1 | 7 | 4839 | |||
XM_017021909.1 | XP_016877398.1 | 7 | 4839 | |||
XM_017021910.1 | XP_016877399.1 | 5 | 4527 | |||
GenBank mRNA | AB209535.1 | - | 1 | 73 | ||
AK091692.1 | - | 2 | 981 | |||
AK225680.1 | - | 2 | 101 | |||
AK291983.1 | - | 2 | 103 | |||
AK293769.1 | - | 2 | 122 | |||
AK301689.1 | - | 2 | 103 | |||
BC002555.2 | - | 2 | 103 | 3138580 | ||
BC006103.1 | - | 2 | 121 | 3957357 | ||
BC019881.1 | - | 2 | 114 | 5114635 | ||
BI459843.1 | - | 2 | 478 | |||
BT006993.1 | - | 1 | 76 | |||
CR933693.1 | - | 2 | 111 | |||
L29217.1 | - | 2 | 132 | |||
L29220.1 | - | 2 | 132 |