Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PXN SIRNA Assays ›
Gene Symbol:
PXN
Gene Name:
paxillin
Gene Aliases:
-
Chromosome Location:
Chr.12: 120210439 - 120265771 on Build GRCh38
UniGene ID:
Hs.446336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
12346
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PXN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080855.2
NP_001074324.1
12
2909
NM_001243756.1
NP_001230685.1
11
2951
NM_002859.3
NP_002850.2
11
2807
NM_025157.4
NP_079433.3
11
2931
XM_005253917.3
XP_005253974.1
7
2306
XM_006719532.1
XP_006719595.1
15
4492
XM_006719534.1
XP_006719597.1
15
4647
XM_006719535.3
XP_006719598.1
16
4575
XM_006719536.2
XP_006719599.1
15
4464
XM_011538622.1
XP_011536924.1
16
4574
XM_011538623.1
XP_011536925.1
16
4761
XM_017019726.1
XP_016875215.1
15
5060
XM_017019727.1
XP_016875216.1
15
4860
XM_017019728.1
XP_016875217.1
15
4851
XM_017019729.1
XP_016875218.1
15
4916
XM_017019731.1
XP_016875220.1
16
4945
XM_017019733.1
XP_016875222.1
13
4475
XM_017019734.1
XP_016875223.1
15
4372
XM_017019735.1
XP_016875224.1
12
4229
XM_017019736.1
XP_016875225.1
13
3899
XM_017019737.1
XP_016875226.1
13
3755
XM_017019738.1
XP_016875227.1
12
3590
XM_017019739.1
XP_016875228.1
11
3488
XM_017019740.1
XP_016875229.1
12
3446
XM_017019741.1
XP_016875230.1
12
3237
XM_017019742.1
XP_016875231.1
11
3344
XM_017019743.1
XP_016875232.1
12
3045
GenBank mRNA
AB209034.1
-
14
4585
AK122964.1
-
12
3237
AK128712.1
-
9
3257
BC003647.2
-
4
1081
BC006840.1
-
1
637
BC010498.1
-
1
845
BC015217.1
-
1
712
BX648777.1
-
11
2924
U14588.1
-
11
2729
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
cell adhesion
cell-matrix adhesion
signal transduction
signal complex assembly
epidermal growth factor receptor signaling pathway
transforming growth factor beta receptor signaling pathway
cellular response to reactive oxygen species
vascular endothelial growth factor receptor signaling pathway
growth hormone receptor signaling pathway
integrin binding
protein binding
beta-catenin binding
zinc ion binding
vinculin binding
protein kinase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz5f2:80/100.66.72.225:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0