Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184891.1 | NP_001171820.1 | 5, 6 | 1315 | ||
NM_025207.4 | NP_079483.3 | 4, 5 | 1759 | |||
NM_201398.2 | NP_958800.1 | 5, 6 | 1315 | |||
GenBank mRNA | AF481877.1 | - | 4, 5 | 1067 | ||
AF520568.1 | - | 4, 5 | 1680 | |||
AK025979.1 | - | 5, 6 | 1243 | |||
BC011378.1 | - | 5, 6 | 1244 | 3855223 | ||
BC020253.1 | - | 3, 4 | 594 | |||
BC021096.1 | - | 5, 6 | 1233 | 4891331 | ||
BC032323.1 | - | 5, 6 | 1250 | 5242727 | ||
DQ458779.1 | - | 4, 5 | 1358 | |||
U79241.1 | - | 3, 4 | 910 |