Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: SASS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304829.1 | NP_001291758.1 | 4 | 617 | ||
NM_194292.2 | NP_919268.1 | 5 | 697 | |||
XM_017000486.1 | XP_016855975.1 | 5 | 463 | |||
GenBank mRNA | AK302513.1 | - | 6 | 447 | ||
AK307824.1 | - | 4 | 484 | |||
AY359522.1 | - | 5 | 557 | |||
BC054889.1 | - | 5 | 560 | |||
BC101026.3 | - | 4 | 410 | 40007432 | ||
BX641109.1 | - | 5 | 516 | |||
BX648406.1 | - | 5 | 453 |