Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NHLRC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198514.3 | NP_940916.2 | 11 | 2412 | ||
GenBank mRNA | AK090631.1 | - | 11 | 2383 | ||
AK098487.1 | - | 9 | 1526 | |||
AL832090.1 | - | 2 | 383 | |||
BC032598.1 | - | 7 | 1176 | 5529619 | ||
BX538287.1 | - | 7 | 1206 | |||
BX647641.1 | - | 9 | 1783 |