Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: GNL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014366.4 | NP_055181.3 | 3 | 310 | ||
NM_206825.1 | NP_996561.1 | 3 | 407 | |||
NM_206826.1 | NP_996562.1 | 3 | 260 | |||
GenBank mRNA | AF191018.1 | - | 3 | 307 | ||
AI028402.1 | - | 3 | 310 | |||
AK027514.1 | - | 3 | 311 | |||
AK027516.1 | - | 3 | 160 | |||
AK315484.1 | - | 3 | 223 | |||
AY825265.1 | - | 2 | 137 | |||
BC001024.2 | - | 3 | 158 | 3342771 | ||
BF732788.1 | - | 3 | 260 | |||
CF272475.1 | - | 3 | 179 |