Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANSL3 SIRNA Assays ›
Gene Symbol:
KANSL3
Gene Name:
KAT8 regulatory NSL complex subunit 3
Gene Aliases:
KIAA1310, NSL3, Rcd1
Chromosome Location:
Chr.2: 96593155 - 96638379 on Build GRCh38
UniGene ID:
Hs.516341
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
125088
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANSL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001115016.2
NP_001108488.1
6
826
NR_047653.1
6
887
NR_047654.1
6
887
NR_047655.1
5
735
NR_047656.1
5
735
NR_047657.1
6
826
NR_047658.1
6
803
XM_005263982.2
XP_005264039.1
6
825
XM_005263984.1
XP_005264041.1
6
825
XM_005263987.1
XP_005264044.1
6
825
XM_011511454.2
XP_011509756.1
6
825
XM_011511455.2
XP_011509757.1
6
794
XM_011511456.2
XP_011509758.1
6
828
XM_011511457.2
XP_011509759.1
6
747
XM_011511458.2
XP_011509760.1
6
825
XM_011511459.2
XP_011509761.1
6
825
XM_011511460.2
XP_011509762.1
6
825
XM_011511461.2
XP_011509763.1
6
825
XM_011511462.2
XP_011509764.1
5
654
XM_011511463.2
XP_011509765.1
6
838
XM_011511464.2
XP_011509766.1
6
897
XM_011511466.2
XP_011509768.1
6
838
XM_011511467.2
XP_011509769.1
5
680
XM_017004470.1
XP_016859959.1
6
825
XM_017004471.1
XP_016859960.1
5
732
XM_017004472.1
XP_016859961.1
6
838
XM_017004473.1
XP_016859962.1
6
838
XM_017004474.1
XP_016859963.1
6
733
GenBank mRNA
AB037731.1
-
6
778
AF311326.1
-
8
639
AK000943.1
-
4
727
AK023813.1
-
6
854
AK302646.1
-
5
676
AK304011.1
-
5
735
AK311617.1
-
5
668
AK314367.1
-
6
763
AY050169.1
-
6
854
BC015469.2
-
7
820
BC024276.1
-
6
763
BC032746.1
-
6
897
BC051763.1
-
4
457
BC063792.1
-
6
756
6504387
CB123459.1
-
4
359
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
histone H4-K5 acetylation
histone H4-K8 acetylation
histone H4-K16 acetylation
histone acetyltransferase activity (H4-K5 specific)
histone acetyltransferase activity (H4-K8 specific)
histone acetyltransferase activity (H4-K16 specific)
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0