Search Thermo Fisher Scientific
Gene Symbol: C22orf29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024627.5 | NP_078903.3 | 3 | 4695 | ||
GenBank mRNA | AK026924.1 | - | 1 | 642 | ||
BX640785.1 | - | 2 | 4784 | |||
BX640969.1 | - | 3 | 4663 | |||
BX640998.1 | - | 3 | 4663 |