Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321118.1 | NP_001308047.1 | 5 | 1180 | ||
NM_001321119.1 | NP_001308048.1 | 5 | 702 | |||
NM_032357.3 | NP_115733.2 | 5 | 1001 | |||
NR_104471.1 | 6 | 1383 | ||||
NR_135548.1 | 6 | 1204 | ||||
GenBank mRNA | AK054693.1 | - | 5 | 967 | ||
AK092196.1 | - | 1 | 1704 | |||
AK124405.1 | - | 2 | 2889 | |||
AK295922.1 | - | 6 | 1040 | |||
BC006429.1 | - | 5 | 763 | 3350642 |