Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNAP25 SIRNA Assays ›
          Gene Symbol:
          SNAP25
          Gene Name:
          synaptosome associated protein 25
          Gene Aliases:
          CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2
          Chromosome Location:
          Chr.20: 10218694 - 10307420 on Build GRCh38
          UniGene ID:
          Hs.167317
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 12589

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNAP25
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001322902.1 NP_001309831.1 6, 7 689
          NM_001322903.1 NP_001309832.1 7, 8 785
          NM_001322904.1 NP_001309833.1 7, 8 755
          NM_001322905.1 NP_001309834.1 7, 8 718
          NM_001322906.1 NP_001309835.1 7, 8 712
          NM_001322907.1 NP_001309836.1 8, 9 913
          NM_001322908.1 NP_001309837.1 8, 9 836
          NM_001322909.1 NP_001309838.1 7, 8 703
          NM_001322910.1 NP_001309839.1 7, 8 810
          NM_003081.4 NP_003072.2 6, 7 658
          NM_130811.3 NP_570824.1 6, 7 658
          XM_005260808.4 XP_005260865.1 6, 7 736
          XM_017028021.1 XP_016883510.1 7, 8 588
          XM_017028022.1 XP_016883511.1 7, 8 680
          GenBank mRNA AK026732.1 - 1, 2 829
          AK090857.1 - 1, 2 1626
          AK094560.1 - 7, 8 744
          AK098767.1 - 6, 7 528
          AK223617.1 - 6, 7 620
          AK289647.1 - 8, 9 819
          AK314359.1 - 6, 7 569
          BC010647.1 - 6, 7 547 3867544
          BI552777.1 - 6, 7 631
          BI602062.1 - 6, 7 635
          BI604120.1 - 6, 7 637
          BT019684.1 - 5, 6 415
          BX647443.1 - 6, 7 530
          D21267.1 - 6, 7 620
          L19760.1 - 6, 7 503
          L19761.1 - 6, 7 503

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001291056, NM_011428, BC018249, AK078038, AF483517, and others.

          Panther Classification:

          Molecular Function -

          SNARE protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neurotransmitter uptake
          chemical synaptic transmission
          neurotransmitter secretion
          locomotory behavior
          associative learning
          regulation of neuron projection development
          glutamate secretion
          synaptic vesicle exocytosis
          synaptic vesicle docking
          synaptic vesicle priming
          synaptic vesicle fusion to presynaptic active zone membrane
          regulation of insulin secretion
          long-term synaptic potentiation
          SNAP receptor activity
          protein binding
          syntaxin-1 binding
          calcium-dependent protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7d94m:80/100.66.78.9:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0