siRNA Details
Gene Symbol: CFAP65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194302.3 | NP_919278.2 | 16, 17 | 2931 | ||
XM_011510903.1 | XP_011509205.1 | 16, 17 | 2992 | |||
XM_011510904.2 | XP_011509206.1 | 16, 17 | 4576 | |||
XM_011510908.1 | XP_011509210.1 | 16, 17 | 2898 | |||
XM_011510909.1 | XP_011509211.1 | 16, 17 | 2938 | |||
XM_011510910.1 | XP_011509212.1 | 16, 17 | 2992 | |||
XM_011510911.1 | XP_011509213.1 | 16, 17 | 2843 | |||
XM_011510912.1 | XP_011509214.1 | 16, 17 | 2886 | |||
XM_011510914.1 | XP_011509216.1 | 15, 16 | 2783 | |||
XM_011510915.1 | XP_011509217.1 | 15, 16 | 2745 | |||
XM_011510916.1 | XP_011509218.1 | 15, 16 | 2713 | |||
XM_011510919.1 | XP_011509221.1 | 16, 17 | 2992 | |||
XM_011510921.1 | XP_011509223.1 | 6, 7 | 1448 | |||
XM_017003752.1 | XP_016859241.1 | 17, 18 | 3059 | |||
XM_017003753.1 | XP_016859242.1 | 16, 17 | 4416 | |||
XM_017003754.1 | XP_016859243.1 | 15, 16 | 2837 | |||
XM_017003755.1 | XP_016859244.1 | 15, 16 | 4421 | |||
GenBank mRNA | AK125957.1 | - | 4, 5 | 720 | ||
AK127189.1 | - | 16, 17 | 2896 | |||
BC130281.1 | - | 4, 5 | 710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
