Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: C19orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198534.2 | NP_940936.2 | 7, 8 | 1351 | ||
XM_005272473.3 | XP_005272530.1 | 7, 8 | 1522 | |||
XM_011527993.1 | XP_011526295.1 | 7, 8 | 1276 | |||
GenBank mRNA | AK093103.1 | - | 7, 8 | 1336 | ||
BC029824.1 | - | 7, 8 | 1311 | 5170600 | ||
BC034779.1 | - | 7, 8 | 1300 | 5168945 |