Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WDR41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018268.3 | NP_060738.2 | 13 | 1969 | ||
XM_005248551.4 | XP_005248608.1 | 11 | 1762 | |||
XM_011543505.2 | XP_011541807.1 | 13 | 1731 | |||
XM_017009606.1 | XP_016865095.1 | 13 | 2379 | |||
GenBank mRNA | AF115511.1 | - | 13 | 1716 | ||
AK001766.1 | - | 13 | 1915 | |||
AK021855.1 | - | 13 | 1755 | |||
AK023977.1 | - | 13 | 1671 | |||
AK293316.1 | - | 11 | 1454 | |||
AL834138.1 | - | 7 | 2650 | |||
BC040241.1 | - | 13 | 1721 | 3447269 |