Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RBM41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171080.1 | NP_001164551.1 | 6 | 1071 | ||
NM_001324242.1 | NP_001311171.1 | 7 | 1143 | |||
NM_001324243.1 | NP_001311172.1 | 7 | 1236 | |||
NM_001324244.1 | NP_001311173.1 | 7 | 1143 | |||
NM_018301.4 | NP_060771.3 | 6 | 1071 | |||
NR_136728.1 | 6 | 739 | ||||
NR_136729.1 | 7 | 833 | ||||
NR_136730.1 | 7 | 1867 | ||||
NR_136731.1 | 7 | 937 | ||||
NR_136732.1 | 6 | 832 | ||||
XM_011530980.2 | XP_011529282.1 | 7 | 1235 | |||
XM_011530981.2 | XP_011529283.1 | 6 | 1163 | |||
GenBank mRNA | AK001878.1 | - | 6 | 1044 | ||
AK023732.1 | - | 7 | 1862 | |||
BC006986.1 | - | 6 | 1048 | 3995903 |