Search
Search

TARS| Gene Symbol: TARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258437.1 | NP_001245366.1 | 3 | 257 | ||
| NM_001258438.1 | NP_001245367.1 | 2 | 431 | |||
| NM_152295.4 | NP_689508.3 | 2 | 431 | |||
| NR_047676.1 | 2 | 431 | ||||
| NR_047677.1 | 2 | 431 | ||||
| NR_047678.1 | 2 | 431 | ||||
| GenBank mRNA | AK095852.1 | - | 2 | 243 | ||
| AK222804.1 | - | 2 | 188 | |||
| AK292346.1 | - | 3 | 257 | |||
| AK293620.1 | - | 2 | 242 | |||
| AK296783.1 | - | 2 | 230 | |||
| AK308342.1 | - | 2 | 214 | |||
| AK311361.1 | - | 242 | ||||
| AK315713.1 | - | 2 | 242 | |||
| BC000517.2 | - | 2 | 200 | 2822851 | ||
| BC010578.1 | - | 2 | 218 | 3458845 | ||
| BC088355.1 | - | 2 | 209 | |||
| M63180.1 | - | 2 | 222 | |||