Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CKMT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020990.4 | NP_066270.1 | 3, 4 | 945 | ||
NR_135748.1 | 2, 3 | 994 | ||||
NR_135749.1 | 2, 3 | 1018 | ||||
NR_135750.1 | 3, 4 | 945 | ||||
NR_135751.1 | 2, 3 | 834 | ||||
XM_005254150.3 | XP_005254207.1 | 2, 3 | 467 | |||
XM_011521194.1 | XP_011519496.1 | 3, 4 | 1033 | |||
XM_011521195.2 | XP_011519497.1 | 3, 4 | 772 | |||
XM_011521196.1 | XP_011519498.1 | 3, 4 | 698 | |||
XM_011521197.2 | XP_011519499.1 | 2, 3 | 532 | |||
XM_011521198.1 | XP_011519500.1 | 3, 4 | 1217 | |||
XM_011521199.2 | XP_011519501.2 | 3, 4 | 1033 | |||
XM_017021902.1 | XP_016877391.1 | 3, 4 | 1217 | |||
GenBank mRNA | AK094322.1 | - | 3, 4 | 523 | ||
AK293685.1 | - | 2, 3 | 261 | |||
AK293790.1 | - | 2, 3 | 801 | |||
AK293939.1 | - | 2, 3 | 598 | |||
AK293961.1 | - | 1, 2 | 383 | |||
AK294002.1 | - | 2, 3 | 643 | |||
AK294063.1 | - | 2, 3 | 416 | |||
AK294735.1 | - | 3, 4 | 416 | |||
AK294908.1 | - | 2, 3 | 833 | |||
AK296271.1 | - | 1, 2 | 78 | |||
AK296472.1 | - | 2, 3 | 480 | |||
AK308770.1 | - | 1, 2 | 350 | |||
BC108652.1 | - | 2, 3 | 468 | 6190035 | ||
BC121001.1 | - | 2, 3 | 532 | 40116549 | ||
BC121002.1 | - | 2, 3 | 532 | 40116552 |
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015001.2 | NP_001015001.1 | 3, 4 | 848 | ||
NM_001321926.1 | NP_001308855.1 | 2, 3 | 716 | |||
NM_001321927.1 | NP_001308856.1 | 3, 4 | 941 | |||
NM_001321928.1 | NP_001308857.1 | 3, 4 | 702 | |||
NM_001321929.1 | NP_001308858.1 | 2, 3 | 744 | |||
NR_135856.1 | 2, 3 | 993 | ||||
XM_005254498.3 | XP_005254555.1 | 2, 3 | 467 | |||
XM_017022369.1 | XP_016877858.1 | 3, 4 | 773 | |||
XM_017022370.1 | XP_016877859.1 | 3, 4 | 698 | |||
XM_017022371.1 | XP_016877860.1 | 3, 4 | 1083 | |||
GenBank mRNA | AK223365.1 | - | 2, 3 | 528 | ||
AK295776.1 | - | 2, 3 | 592 | |||
AK316052.1 | - | 3, 4 | 392 | |||
AK316124.1 | - | 3, 4 | 799 | |||
AK316319.1 | - | 3, 4 | 702 | |||
BC001926.1 | - | 2, 3 | 533 | 2820642 | ||
BC006467.1 | - | 2, 3 | 459 | 3535486 | ||
BM923431.1 | - | 3, 4 | 767 | |||
BT006628.1 | - | 2, 3 | 356 |