Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TGFB2 SIRNA Assays ›
          Gene Symbol:
          TGFB2
          Gene Name:
          transforming growth factor beta 2
          Gene Aliases:
          LDS4, TGF-beta2
          Chromosome Location:
          Chr.1: 218345334 - 218444619 on Build GRCh38
          UniGene ID:
          Hs.133379
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 12748

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TGFB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135599.2 NP_001129071.1 1 1574
          NM_003238.3 NP_003229.1 1 1574
          GenBank mRNA AB209842.1 - 1 1502
          AK295671.1 - 1 53
          AK296504.1 - 1 207
          AK313106.1 - 1 311
          BC096235.4 - 1 360 40005424
          BC096236.4 - 1 364
          BC099635.1 - 1 364 40005426
          M19154.1 - 1 673
          Y00083.1 - 1 387

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031131, XM_006250448, BC100663, AJ132718, AF153013, and others.
          This siRNA also targets the following transcripts in Mouse: NM_009367, BC011170, AK141552, X57413, AJ639842.

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          cell morphogenesis
          cartilage condensation
          angiogenesis
          eye development
          response to hypoxia
          epithelial to mesenchymal transition
          hair follicle development
          blood vessel remodeling
          platelet degranulation
          heart morphogenesis
          outflow tract septum morphogenesis
          membranous septum morphogenesis
          heart valve morphogenesis
          atrioventricular valve morphogenesis
          pulmonary valve morphogenesis
          endocardial cushion morphogenesis
          cardiac right ventricle morphogenesis
          ventricular trabecula myocardium morphogenesis
          endocardial cushion fusion
          atrial septum primum morphogenesis
          protein phosphorylation
          cell cycle arrest
          transforming growth factor beta receptor signaling pathway
          SMAD protein import into nucleus
          cell-cell signaling
          axon guidance
          salivary gland morphogenesis
          heart development
          cell death
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          glial cell migration
          response to wounding
          embryo development
          cardioblast differentiation
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial cell migration
          negative regulation of alkaline phosphatase activity
          positive regulation of epithelial to mesenchymal transition
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of macrophage cytokine production
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell growth
          cell migration
          signal transduction by protein phosphorylation
          hemopoiesis
          collagen fibril organization
          positive regulation of cell growth
          negative regulation of cell growth
          neutrophil chemotaxis
          hair follicle morphogenesis
          activation of protein kinase activity
          response to progesterone
          positive regulation of stress-activated MAPK cascade
          regulation of transforming growth factor beta2 production
          regulation of actin cytoskeleton organization
          positive regulation of cell adhesion mediated by integrin
          ascending aorta morphogenesis
          wound healing
          dopamine biosynthetic process
          odontogenesis
          response to drug
          catagen
          uterine wall breakdown
          positive regulation of neuron apoptotic process
          cell-cell junction organization
          positive regulation of integrin biosynthetic process
          positive regulation of ossification
          positive regulation of cell cycle
          positive regulation of heart contraction
          negative regulation of Ras protein signal transduction
          somatic stem cell division
          embryonic digestive tract development
          neuron fate commitment
          neuron development
          generation of neurons
          negative regulation of epithelial cell proliferation
          positive regulation of protein secretion
          negative regulation of immune response
          positive regulation of immune response
          positive regulation of cell division
          positive regulation of catagen
          positive regulation of cardioblast differentiation
          cardiac muscle cell proliferation
          cardiac epithelial to mesenchymal transition
          face morphogenesis
          pathway-restricted SMAD protein phosphorylation
          SMAD protein signal transduction
          ventricular septum morphogenesis
          atrial septum morphogenesis
          negative regulation of cartilage development
          pharyngeal arch artery morphogenesis
          positive regulation of activation-induced cell death of T cells
          positive regulation of extracellular matrix disassembly
          extrinsic apoptotic signaling pathway
          positive regulation of protein localization to nucleus
          regulation of apoptotic process involved in outflow tract morphogenesis
          regulation of complement-dependent cytotoxicity
          positive regulation of GTP binding
          negative regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of epithelial to mesenchymal transition involved in endocardial cushion formation
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          beta-amyloid binding
          receptor signaling protein serine/threonine kinase activity
          receptor binding
          type II transforming growth factor beta receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          protein binding
          growth factor activity
          type III transforming growth factor beta receptor binding
          protein homodimerization activity
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0