Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CXorf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025159.2 | NP_079435.1 | 3 | 1433 | ||
GenBank mRNA | AK021639.1 | - | 3 | 1314 | ||
BC020611.1 | - | 3 | 1361 | 4768735 |