Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TPK1 SIRNA Assays ›
          Gene Symbol:
          TPK1
          Gene Name:
          thiamin pyrophosphokinase 1
          Gene Aliases:
          HTPK1, PP20, THMD5
          Chromosome Location:
          Chr.7: 144451941 - 144836424 on Build GRCh38
          UniGene ID:
          Hs.660232
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 1279

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TPK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042482.1 NP_001035947.1 4, 5 283
          NM_022445.3 NP_071890.2 4, 5 283
          XM_005249970.1 XP_005250027.1 4, 5 261
          XM_011516031.1 XP_011514333.1 4, 5 284
          XM_011516032.2 XP_011514334.1 4, 5 420
          XM_011516033.2 XP_011514335.1 4, 5 290
          XM_011516034.2 XP_011514336.1 4, 5 347
          XM_011516035.2 XP_011514337.1 4, 5 284
          XM_011516040.2 XP_011514342.1 4, 5 284
          XM_011516043.1 XP_011514345.1 4, 5 284
          XM_011516046.1 XP_011514348.1 4, 5 284
          XM_017011969.1 XP_016867458.1 4, 5 284
          XM_017011972.1 XP_016867461.1 4, 5 284
          XM_017011973.1 XP_016867462.1 4, 5 428
          XM_017011976.1 XP_016867465.1 4, 5 283
          XM_017011979.1 XP_016867468.1 4, 5 415
          XM_017011982.1 XP_016867471.1 2, 3 182
          XM_017011986.1 XP_016867475.1 1, 2 69
          GenBank mRNA AB028138.1 - 3, 4 198
          AF297710.1 - 4, 5 270
          AK026374.1 - 2, 3 90
          AK091736.1 - 4, 5 222
          AK289652.1 - 4, 5 258
          AK308693.1 - 4, 5 283
          AY206415.1 - 4, 5 265
          BC068460.1 - 4, 5 262 5288626
          BI770816.1 - 3, 4 199
          CR749248.1 - 2, 3 270
          DA135998.1 - 4, 5 283

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          thiamine metabolic process
          thiamine diphosphate biosynthetic process
          phosphorylation
          thiamine-containing compound metabolic process
          thiamine diphosphokinase activity
          ATP binding
          kinase activity
          thiamine binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0