Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC38A9 SIRNA Assays ›
          Gene Symbol:
          SLC38A9
          Gene Name:
          solute carrier family 38 member 9
          Gene Aliases:
          URLC11
          Chromosome Location:
          Chr.5: 55625845 - 55712344 on Build GRCh38
          UniGene ID:
          Hs.742101
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 128221

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC38A9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282429.1 NP_001269358.1 3 299
          NM_173514.3 NP_775785.2 3 299
          XM_006714537.2 XP_006714600.1 5 533
          XM_006714538.3 XP_006714601.1 4 420
          XM_006714539.3 XP_006714602.1 4 463
          XM_011543173.1 XP_011541475.1 5 521
          XM_011543174.1 XP_011541476.1 6 619
          XM_011543176.1 XP_011541478.1 5 533
          XM_011543177.1 XP_011541479.1 5 533
          XM_011543178.1 XP_011541480.1 5 533
          XM_011543180.1 XP_011541482.1 5 533
          XM_011543182.1 XP_011541484.1 5 533
          XM_011543183.1 XP_011541485.1 5 533
          XM_011543184.1 XP_011541486.1 5 533
          XM_017009077.1 XP_016864566.1 3 231
          XM_017009078.1 XP_016864567.1 3 308
          XM_017009079.1 XP_016864568.1 3 231
          XM_017009080.1 XP_016864569.1 4 420
          XM_017009081.1 XP_016864570.1 3 231
          XM_017009082.1 XP_016864571.1 3 231
          GenBank mRNA AB105188.1 - 3 231
          AK075190.1 - 3 231
          AK122957.1 - 3 279
          AK128743.1 - 804
          BC066891.1 - 3 288 4373965
          BC101362.2 - 3 230 40025719
          DC371481.1 - 3 299

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          amino acid transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          amino acid transmembrane transport
          amino acid transport
          positive regulation of TOR signaling
          cellular response to amino acid stimulus
          protein binding
          amino acid transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0