Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: THBS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003247.3 | NP_003238.2 | 14 | 2287 | ||
GenBank mRNA | AA853383.1 | - | 2 | 81 | ||
AK292429.1 | - | 14 | 2244 | |||
BC146676.1 | - | 13 | 2094 | 40148868 | ||
BC150175.1 | - | 13 | 2098 | 40147958 | ||
L12350.1 | - | 14 | 2206 | |||
M81339.1 | - | 3 | 291 |