Search Thermo Fisher Scientific
- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003818.2 | NP_001003818.1 | 8 | 1576 | ||
NM_001198644.1 | NP_001185573.1 | 7 | 1086 | |||
NM_001198645.1 | NP_001185574.1 | 6 | 990 | |||
NM_058166.4 | NP_477514.1 | 8 | 1377 | |||
GenBank mRNA | AF220030.1 | - | 8 | 1365 | ||
AK023210.1 | - | 5 | 628 | |||
AK027664.1 | - | 8 | 1369 | |||
AK290172.1 | - | 8 | 1576 | |||
AK293295.1 | - | 6 | 819 | |||
AK298301.1 | - | 7 | 915 | |||
AK316178.1 | - | 8 | 1043 | |||
AK316289.1 | - | 8 | 1171 | |||
BC047564.1 | - | 8 | 1555 | 4825118 | ||
BC065575.1 | - | 8 | 1102 | 4609084 | ||
BG776117.1 | - | 194 | ||||
CR749260.1 | - | 9 | 1272 |