Search
Search

TARS| Gene Symbol: TARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258437.1 | NP_001245366.1 | 14 | 1577 | ||
| NM_001258438.1 | NP_001245367.1 | 14 | 1850 | |||
| NM_152295.4 | NP_689508.3 | 13 | 1751 | |||
| NR_047676.1 | 14 | 1872 | ||||
| NR_047677.1 | 12 | 1590 | ||||
| NR_047678.1 | 12 | 1560 | ||||
| GenBank mRNA | AK095852.1 | - | 12 | 1372 | ||
| AK222804.1 | - | 13 | 1508 | |||
| AK292346.1 | - | 14 | 1577 | |||
| AK293620.1 | - | 14 | 1661 | |||
| AK296783.1 | - | 12 | 1389 | |||
| AK308342.1 | - | 14 | 1655 | |||
| AK315713.1 | - | 13 | 1562 | |||
| BC000517.2 | - | 13 | 1520 | 2822851 | ||
| BC010578.1 | - | 13 | 1538 | 3458845 | ||
| BC088355.1 | - | 13 | 1529 | |||
| M63180.1 | - | 13 | 1542 | |||