Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TAS2R60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177437.1 | NP_803186.1 | 1 | 728 | ||
GenBank mRNA | BC100938.2 | - | 1 | 728 | 40004056 | |
BC100939.2 | - | 1 | 728 | 40004057 | ||
BC100940.1 | - | 1 | 728 | 40004058 |