Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SUMO1 SIRNA Assays ›
          Gene Symbol:
          SUMO1 SUMO1P3
          Gene Name:
          small ubiquitin-like modifier 1
          SUMO1 pseudogene 3
          Gene Aliases:
          DAP1, GMP1, OFC10, PIC1, SENP2, SMT3, SMT3C, SMT3H3, UBL1
          -
          Chromosome Location:
          Chr.2: 202206180 - 202238599 on Build GRCh38
          Chromosome Location:
          Chr.1: 160317265 - 160318474 on Build GRCh38
          UniGene ID:
          Hs.81424
          Hs.621179
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 12999

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SUMO1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005781.1 NP_001005781.1 2, 3 243
          NM_001005782.1 NP_001005782.1 1, 2 168
          NM_003352.4 NP_003343.1 2, 3 243
          GenBank mRNA AB062294.1 - 2, 3 219
          AK311840.1 - 2, 3 208
          BC005899.1 - 2, 3 210
          BC006462.1 - 2, 3 226 3546125
          BC053528.1 - 2, 3 201 3452929
          BC066306.1 - 2, 3 219 4818792
          BG503431.1 - 2, 3 246
          BG569821.1 - 1, 2 136
          BT006632.1 - 1 95
          BU532734.1 - 192
          CR542147.1 - 1 95
          CR542156.1 - 1 95
          U38784.1 - 3, 4 161
          U61397.1 - 2, 3 240
          U67122.1 - 2, 3 175
          U72722.1 - 1 95
          U83117.1 - 2, 3 230
          X99586.1 - 136
          Gene Symbol: SUMO1P3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002190.1 1 231

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          double-strand break repair via nonhomologous end joining
          viral process
          protein sumoylation
          PML body organization
          positive regulation of protein complex assembly
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          regulation of protein localization
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          negative regulation of action potential
          negative regulation of transcription, DNA-templated
          protein stabilization
          palate development
          regulation of interferon-gamma-mediated signaling pathway
          global genome nucleotide-excision repair
          regulation of cardiac muscle cell contraction
          protein localization to nuclear pore
          positive regulation of calcium-transporting ATPase activity
          negative regulation of delayed rectifier potassium channel activity
          protein binding
          transcription factor binding
          potassium channel regulator activity
          protein tag
          ubiquitin protein ligase binding
          ion channel binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0