Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BCL9L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182557.2 | NP_872363.1 | 2 | 1228 | ||
XM_005271511.2 | XP_005271568.1 | 4 | 1228 | |||
XM_005271512.2 | XP_005271569.1 | 3 | 678 | |||
XM_005271513.2 | XP_005271570.1 | 2 | 677 | |||
XM_006718815.2 | XP_006718878.1 | 3 | 726 | |||
XM_011542756.2 | XP_011541058.1 | 2 | 1184 | |||
XM_017017572.1 | XP_016873061.1 | 4 | 1477 | |||
XM_017017573.1 | XP_016873062.1 | 3 | 1829 | |||
XM_017017574.1 | XP_016873063.1 | 3 | 1376 | |||
GenBank mRNA | AB094091.1 | - | 2 | 1228 | ||
AY296059.1 | - | 2 | 248 |