Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDRG4 SIRNA Assays ›
          Gene Symbol:
          NDRG4
          Gene Name:
          NDRG family member 4
          Gene Aliases:
          BDM1, SMAP-8, SMAP8
          Chromosome Location:
          Chr.16: 58463645 - 58515403 on Build GRCh38
          UniGene ID:
          Hs.322430
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 130252

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NDRG4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130487.1 NP_001123959.1 16 2693
          NM_001242833.1 NP_001229762.1 15 2392
          NM_001242834.1 NP_001229763.1 14 2356
          NM_001242835.1 NP_001229764.1 15 2341
          NM_001242836.1 NP_001229765.1 14 2302
          NM_020465.3 NP_065198.1 16 2468
          NM_022910.3 NP_075061.1 16 2565
          NR_040072.1 14 2394
          XM_006721252.3 XP_006721315.1 18 2607
          XM_006721253.3 XP_006721316.1 18 2540
          XM_006721254.3 XP_006721317.1 17 2594
          XM_006721256.3 XP_006721319.1 18 2538
          XM_006721257.3 XP_006721320.1 17 2579
          XM_006721258.3 XP_006721321.1 17 2483
          XM_006721259.3 XP_006721322.1 17 2548
          XM_006721260.3 XP_006721323.1 17 2486
          XM_006721261.3 XP_006721324.1 16 2431
          XM_006721262.3 XP_006721325.1 15 2512
          XM_011523290.2 XP_011521592.1 18 2694
          XM_011523291.2 XP_011521593.1 17 2658
          XM_011523292.2 XP_011521594.1 17 2655
          XM_011523293.2 XP_011521595.1 17 2652
          XM_011523294.2 XP_011521596.1 18 2473
          XM_017023582.1 XP_016879071.1 16 2619
          XM_017023584.1 XP_016879073.1 17 2568
          XM_017023585.1 XP_016879074.1 16 2555
          XM_017023586.1 XP_016879075.1 16 2502
          XM_017023587.1 XP_016879076.1 17 2499
          XM_017023588.1 XP_016879077.1 16 2444
          XM_017023589.1 XP_016879078.1 16 2509
          XM_017023590.1 XP_016879079.1 17 2434
          GenBank mRNA AB021172.1 - 16 2368
          AB033006.2 - 15 2327
          AB044944.1 - 14 2302
          AB044945.1 - 15 2341
          AB044946.1 - 15 2315
          AF007138.1 - 3 1426
          AF308608.1 - 14 2309
          AK055038.1 - 14 2304
          AK055148.1 - 1 2042
          AK124005.1 - 5 1347
          AK126574.1 - 16 2465
          AK126729.1 - 18 4454
          AK131248.1 - 14 2356
          AL136584.1 - 14 2288
          BC001527.1 - 14 2286
          BC011795.2 - 14 2290 4139594
          CR936733.1 - 16 2379

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          heart looping
          response to stress
          brain development
          visual learning
          negative regulation of platelet-derived growth factor receptor signaling pathway
          positive regulation of neuron projection development
          negative regulation of smooth muscle cell migration
          cell growth
          cell differentiation
          embryonic heart tube development
          vesicle docking
          negative regulation of smooth muscle cell proliferation
          cardiac muscle cell proliferation
          cell migration involved in heart development
          positive regulation of ERK1 and ERK2 cascade
          regulation of endocytic recycling
          molecular_function
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0