Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DHDDS SIRNA Assays ›
Gene Symbol:
DHDDS
Gene Name:
dehydrodolichyl diphosphate synthase subunit
Gene Aliases:
CIT, CPT, DS, HDS, RP59
Chromosome Location:
Chr.1: 26432282 - 26471306 on Build GRCh38
UniGene ID:
Hs.369385
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130782
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DHDDS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243564.1
NP_001230493.1
7, 8
824
NM_001243565.1
NP_001230494.1
7, 8
809
NM_001319959.1
NP_001306888.1
8, 9
949
NM_024887.3
NP_079163.2
9
929
NM_205861.2
NP_995583.1
8, 9
926
XM_006710912.2
XP_006710975.1
9
988
XM_006710913.2
XP_006710976.1
9
867
XM_006710914.2
XP_006710977.1
9
864
XM_006710915.2
XP_006710978.1
8
799
XM_006710916.2
XP_006710979.1
9
978
XM_006710917.2
XP_006710980.1
9
931
XM_006710918.2
XP_006710981.1
9
887
XM_011542183.2
XP_011540485.1
9
964
XM_011542184.2
XP_011540486.1
8, 9
982
XM_011542185.2
XP_011540487.1
8
883
XM_011542186.2
XP_011540488.1
7, 8
880
XM_017002379.1
XP_016857868.1
8, 9
864
XM_017002380.1
XP_016857869.1
8, 9
961
XM_017002381.1
XP_016857870.1
8, 9
975
XM_017002382.1
XP_016857871.1
8, 9
884
XM_017002383.1
XP_016857872.1
7, 8
873
XM_017002384.1
XP_016857873.1
7, 8
826
GenBank mRNA
AB090852.1
-
7, 8
787
AK023164.1
-
9
859
AK297134.1
-
6, 7
744
AK308187.1
-
8, 9
868
AK316485.1
-
7, 8
767
BC003643.2
-
7, 8
831
3532466
BC004117.2
-
7, 8
751
3843062
BC034152.1
-
2, 3
151
BX648507.1
-
8, 9
951
GU727641.1
-
7, 8
831
Back To Top
More Information
Panther Classification:
Molecular Function -
acyltransferase
Gene Ontology Categories:
Function(s)
Process(es)
protein glycosylation
dolichyl diphosphate biosynthetic process
polyprenyltransferase activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0