Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CEP76 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271989.1 | NP_001258918.1 | 8 | 1225 | ||
NM_024899.3 | NP_079175.2 | 9 | 1450 | |||
NR_073537.1 | 9 | 1445 | ||||
XM_005258149.4 | XP_005258206.1 | 9 | 1429 | |||
XM_017025981.1 | XP_016881470.1 | 9 | 1429 | |||
XM_017025982.1 | XP_016881471.1 | 9 | 1510 | |||
XM_017025983.1 | XP_016881472.1 | 8 | 1300 | |||
XM_017025984.1 | XP_016881473.1 | 9 | 1412 | |||
XM_017025985.1 | XP_016881474.1 | 9 | 1378 | |||
XM_017025986.1 | XP_016881475.1 | 8 | 1187 | |||
XM_017025987.1 | XP_016881476.1 | 8 | 1154 | |||
GenBank mRNA | AK022604.1 | - | 9 | 1312 | ||
AK301964.1 | - | 8 | 1187 | |||
AK303117.1 | - | 8 | 1172 | |||
BC026307.1 | - | 9 | 1377 | 4792211 | ||
BC033026.1 | - | 9 | 1312 |