Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TRIM45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145635.1 | NP_001139107.1 | 6 | 2253 | ||
NM_025188.3 | NP_079464.2 | 6 | 2307 | |||
XM_011542199.2 | XP_011540501.1 | 7 | 1603 | |||
GenBank mRNA | AK023243.1 | - | 6 | 2246 | ||
AK222901.1 | - | 6 | 2251 | |||
AY669488.1 | - | 6 | 2307 | |||
BC034943.2 | - | 6 | 2226 | 4826874 |