Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RIOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031480.2 | NP_113668.2 | 13, 14 | 1451 | ||
NM_153005.1 | NP_694550.1 | 6, 7 | 761 | |||
XM_005249446.3 | XP_005249503.1 | 13, 14 | 1496 | |||
XM_011514933.2 | XP_011513235.2 | 13, 14 | 1413 | |||
GenBank mRNA | AF258661.1 | - | 6, 7 | 761 | ||
AK054568.1 | - | 13, 14 | 1442 | |||
AK314467.1 | - | 13, 14 | 1480 | |||
AL834277.1 | - | 12, 13 | 1150 | |||
BC006104.1 | - | 13, 14 | 1451 | 3537086 |