Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127174.1 | NP_001120646.1 | 6 | 1308 | ||
NM_001127175.1 | NP_001120647.1 | 6 | 1419 | |||
NM_001127176.1 | NP_001120648.1 | 7 | 1575 | |||
NM_031939.3 | NP_114145.2 | 8 | 1678 | |||
XM_005258365.3 | XP_005258422.1 | 7 | 1488 | |||
XM_011526226.2 | XP_011524528.1 | 7 | 1459 | |||
XM_017026035.1 | XP_016881524.1 | 7 | 1455 | |||
XM_017026036.1 | XP_016881525.1 | 7 | 1459 | |||
XM_017026037.1 | XP_016881526.1 | 6 | 1448 | |||
GenBank mRNA | AK054702.1 | - | 8 | 1612 | ||
AK294757.1 | - | 6 | 1265 | |||
AK295666.1 | - | 6 | 1420 |