Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LTV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032860.3 | NP_116249.2 | 8 | 1072 | ||
GenBank mRNA | AK027815.1 | - | 6 | 821 | ||
AK091447.1 | - | 3 | 4121 | |||
BC009855.2 | - | 8 | 1072 | 3926227 |