Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DIXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037954.3 | NP_001033043.1 | 18 | 2098 | ||
NM_033425.4 | NP_219493.1 | 14 | 1631 | |||
XM_017018466.1 | XP_016873955.1 | 19 | 2058 | |||
XM_017018467.1 | XP_016873956.1 | 19 | 1926 | |||
XM_017018468.1 | XP_016873957.1 | 14 | 1242 | |||
XM_017018469.1 | XP_016873958.1 | 15 | 1267 | |||
GenBank mRNA | AB051522.1 | - | 14 | 1538 | ||
AK055469.1 | - | 5 | 410 | |||
AK055899.1 | - | 12 | 1154 | |||
AK289807.1 | - | 14 | 1607 | |||
AK294954.1 | - | 9 | 797 | |||
BC033034.2 | - | 14 | 1590 | 4820652 | ||
BC041626.1 | - | 11 | 825 | |||
DQ642016.1 | - | 18 | 2098 |